Learn More
Genscript Corporation Benz-Neburase His
Supplier: Genscript Corporation Z03626100
This enzyme can be used to remove all forms of DNA and RNA including double stranded single stranded linearized and circular forms