Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzaldehyde, 2-amino-3,5-dibromo | 50910-55-9 | MFCD00671100 | 100.0% | 278.93 | C7H5Br2NO | 100 G
SDP

Catalog No. 5000242363
Encompass_Preferred

2-Amino-3,5-dibromobenzaldehyde (CAS 50910-55-9) is an organic biochemical reagent used in life-science research as an assay reagent, analytical reference, and synthetic intermediate. It is supplied as a solid, light yellow to yellow material with high purity and defined storage conditions to preserve stability.

  • High purity (99.98%) suitable for analytical and synthetic use.
  • Solid, light yellow to yellow appearance for easy handling.
  • Molecular formula C7H5Br2NO; molecular weight 278.93.
  • Suitable as a biochemical assay reagent and synthetic intermediate.
  • Recommended storage at -20°C under nitrogen; in solvent: -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-002-42363 Supplier Medchemexpress LLC Supplier No. HY107849100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Amino-3,5-dibromobenzaldehyde (CAS 50910-55-9) is an organic biochemical reagent used in life-science research as an assay reagent, analytical reference, and synthetic intermediate. It is supplied as a solid, light yellow to yellow material with high purity and defined storage conditions to preserve stability.

  • High purity (99.98%) suitable for analytical and synthetic use.
  • Solid, light yellow to yellow appearance for easy handling.
  • Molecular formula C7H5Br2NO; molecular weight 278.93.
  • Suitable as a biochemical assay reagent and synthetic intermediate.
  • Recommended storage at -20°C under nitrogen; in solvent: -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.