Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzaldehyde, 2-amino-3,5-dibromo- | 50910-55-9 | MFCD00671100 | 100.0% | 278.93 g/mol | C7H5Br2NO | 50 G
SDP

Catalog No. 5000242275
Encompass_Preferred

2-Amino-3,5-dibromobenzaldehyde is an aromatic aldehyde used as a biochemical reagent and a synthetic intermediate in small-molecule and organic synthesis research. It is typically supplied as a light yellow solid and should be handled and stored under inert atmosphere at low temperature to preserve stability.

  • High purity (listed at 99.98%) suitable for analytical and synthetic use.
  • Molecular formula C7H5Br2NO; molecular weight 278.93 g/mol.
  • Available in multiple pack sizes to support small-scale and larger preparations.
  • Solid form with light yellow to yellow appearance.
  • Storage under nitrogen at -20°C recommended; solutions stable at -80°C for extended storage.
  • Datasheet, certificate of analysis, and safety data sheet available for quality and handling guidance.

Catalog No. 50-002-42275 Supplier Medchemexpress LLC Supplier No. HY10784950G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Amino-3,5-dibromobenzaldehyde is an aromatic aldehyde used as a biochemical reagent and a synthetic intermediate in small-molecule and organic synthesis research. It is typically supplied as a light yellow solid and should be handled and stored under inert atmosphere at low temperature to preserve stability.

  • High purity (listed at 99.98%) suitable for analytical and synthetic use.
  • Molecular formula C7H5Br2NO; molecular weight 278.93 g/mol.
  • Available in multiple pack sizes to support small-scale and larger preparations.
  • Solid form with light yellow to yellow appearance.
  • Storage under nitrogen at -20°C recommended; solutions stable at -80°C for extended storage.
  • Datasheet, certificate of analysis, and safety data sheet available for quality and handling guidance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.