Learn More
Medchemexpress LLC Benzaldehyde, 2-chloro-6-fluoro- | 387-45-1 | MFCD00003306 | 99.7% | 158.56 g/mol | C7H4ClFO | 500 G
SDP
Catalog No. 5000302438
Encompass_Preferred
2-Chloro-6-fluorobenzaldehyde is a halogenated benzaldehyde intermediate used in organic synthesis and medicinal chemistry research, supplied as a high-purity reagent for laboratory applications.
  • High purity for reliable synthetic and analytical results (≈99.7%).
  • Suitable as a building block for derivatization and cross-coupling reactions.
  • Well-characterized physical properties: C7H4ClFO, 158.56 g/mol; solid below 32°C and liquid above 35°C.
  • Supplied in multiple pack sizes to accommodate small-scale research or larger synthesis needs.
  • Stable under normal storage conditions when kept in a closed container.
Catalog No. 50-003-02438 Supplier Medchemexpress LLC Supplier No. HYW015477500G
2-Chloro-6-fluorobenzaldehyde is a halogenated benzaldehyde intermediate used in organic synthesis and medicinal chemistry research, supplied as a high-purity reagent for laboratory applications.
  • High purity for reliable synthetic and analytical results (≈99.7%).
  • Suitable as a building block for derivatization and cross-coupling reactions.
  • Well-characterized physical properties: C7H4ClFO, 158.56 g/mol; solid below 32°C and liquid above 35°C.
  • Supplied in multiple pack sizes to accommodate small-scale research or larger synthesis needs.
  • Stable under normal storage conditions when kept in a closed container.