Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzaldehyde, 3-methoxy- | 591-31-1 | 136.15 | 25 G
SDP

Catalog No. 5000449275
Encompass_Preferred

m-Anisaldehyde, also known as 3-Methoxybenzaldehyde, is an endogenous metabolite found as a colorless to light yellow liquid with a density of 1.117 g/cm³. It has a chemical formula of C8H8O2 and is associated with thyroid autoimmunity during early pregnancy.

  • Appears as a colorless to light yellow liquid
  • Density of 1.117 g/cm³
  • Chemical formula is C8H8O2
  • Associated with thyroid autoimmunity during early pregnancy
  • Derived from plants
  • Soluble in various solvents including DMSO, PEG300, Tween-80, and Corn Oil
  • Classified as a ketone, aldehyde, and acid

Catalog No. 50-004-49275 Supplier Medchemexpress LLC Supplier No. 25GHYW007346
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-Anisaldehyde, also known as 3-Methoxybenzaldehyde, is an endogenous metabolite found as a colorless to light yellow liquid with a density of 1.117 g/cm³. It has a chemical formula of C8H8O2 and is associated with thyroid autoimmunity during early pregnancy.

  • Appears as a colorless to light yellow liquid
  • Density of 1.117 g/cm³
  • Chemical formula is C8H8O2
  • Associated with thyroid autoimmunity during early pregnancy
  • Derived from plants
  • Soluble in various solvents including DMSO, PEG300, Tween-80, and Corn Oil
  • Classified as a ketone, aldehyde, and acid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.