Learn More
Medchemexpress LLC Benzaldehyde, 4-methyl- | 104-87-0 | 99.16% | 120.15 | 250 G
SDP
Catalog No. 5000451291
Encompass_Preferred
p-Tolualdehyde is an active small molecule functioning as a crucial drug intermediate. It is also utilized in the synthesis of isoxazole derivatives, which show promise as multidrug resistance (MDR) reversal agents, and serves as a model for forming fluorescent compounds for aldehyde determination.
  • Used as a drug intermediate
  • Applied in the synthesis of isoxazole derivatives
  • Potential in developing multidrug resistance reversal agents
  • Forms fluorescent compounds for aldehyde determination
  • Exhibits an IC50 of 70.74 μM against B16-F10 cells
Catalog No. 50-004-51291 Supplier Medchemexpress LLC Supplier No. 250GHYW012860
p-Tolualdehyde is an active small molecule functioning as a crucial drug intermediate. It is also utilized in the synthesis of isoxazole derivatives, which show promise as multidrug resistance (MDR) reversal agents, and serves as a model for forming fluorescent compounds for aldehyde determination.
  • Used as a drug intermediate
  • Applied in the synthesis of isoxazole derivatives
  • Potential in developing multidrug resistance reversal agents
  • Forms fluorescent compounds for aldehyde determination
  • Exhibits an IC50 of 70.74 μM against B16-F10 cells