Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Benzaldehyde dimethyl acetal

Catalog No. 226076500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-607-6500G 500 g
22-607-6100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-607-6500G Supplier Sigma Aldrich Supplier No. 226076500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The abstraction of α-hydrogen atoms from benzaldehyde dimethyl acetal by active bromine at 80C has been investigated.Benzaldehyde dimethyl acetal used as an effective reagent for the construction of selenocarbonyl compounds.

Specifications

CAS 1125-88-8
Density 1.014 g/mL (at 25°C (literature))
Boiling Point 87°C to 89°C (18 mmHg)
Molecular Formula C9H12O2
Refractive Index n20/D 1.493 (literature)
Linear Formula C6H5CH(OCH3)2
MDL Number MFCD00008491
Quantity 500 g
Synonym alpha,alpha-Dimethoxytoluene
Molecular Weight (g/mol) 152.19
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.