Learn More
Medchemexpress LLC Benzaldehyde-PEG4-azide | 1151451-77-2 | 97.0% | 323.34 | 1 G
SDP
Catalog No. 5000365492
Encompass_Preferred
Benzaldehyde-PEG4-azide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a versatile click chemistry reagent due to its Azide group, enabling participation in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules, as well as strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups. PROTACs, which incorporate this linker, are designed to leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Capable of copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Facilitates selective degradation of target proteins via the ubiquitin-proteasome system
Catalog No. 50-003-65492 Supplier Medchemexpress LLC Supplier No. 1GHY133455
Benzaldehyde-PEG4-azide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a versatile click chemistry reagent due to its Azide group, enabling participation in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules, as well as strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups. PROTACs, which incorporate this linker, are designed to leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Capable of copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Facilitates selective degradation of target proteins via the ubiquitin-proteasome system