Learn More
Medchemexpress LLC Benzalkonium chloride | 8001-54-5 | 51% | 50 MG
SDP
Catalog No. 5000410508
Encompass_Preferred
Benzalkonium chloride (51% in water) is a potent antimicrobial agent commonly used as a preservative in eye drops. Research indicates it exhibits dose-dependent cytotoxicity in various cell types, with different effects observed in human keratinocytes, NHEK, and NB1RGB cell cultures. Studies in vivo have shown varying impacts on nasal epithelium in animal models.
  • Potent anti-microbial agent
  • Used as a preservative in eye drops
  • Exhibits dose-dependent cytotoxicity in cells
  • Provided as a 51% solution in water
Catalog No. 50-004-10508 Supplier Medchemexpress LLC Supplier No. HYB223250MG
Benzalkonium chloride (51% in water) is a potent antimicrobial agent commonly used as a preservative in eye drops. Research indicates it exhibits dose-dependent cytotoxicity in various cell types, with different effects observed in human keratinocytes, NHEK, and NB1RGB cell cultures. Studies in vivo have shown varying impacts on nasal epithelium in animal models.
  • Potent anti-microbial agent
  • Used as a preservative in eye drops
  • Exhibits dose-dependent cytotoxicity in cells
  • Provided as a 51% solution in water