Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzalkonium chloride, MP Biomedicals™

Catalog No. ICN15043083 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15043083 250 g
ICN15043091 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15043083 Supplier MP Biomedicals, Inc Supplier No. 0215043083
Add to Cart
Add to Cart

Chemical Identifiers

CAS 8001-54-5
Molecular Formula C9H13ClN
Molecular Weight (g/mol) 170.66
Synonym Ammonyx

Specifications

Color Colorless
Physical Form Viscous Solution
Quantity 250 g
Chemical Name or Material Benzalkonium chloride
Recommended Storage Store at Room Temperature(15°C to 30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.