Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Benzalkonium Chloride Solution

Catalog No. 63249500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-249-500ML 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 63-249-500ML Supplier Sigma Aldrich Supplier No. 63249500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Benzalkonium chloride is a quaternary ammonium salt that can be used as a preservative, antiseptic, and disinfectant in ocular drug products.Benzalkonium chloride (BAC) can be used as: A cationic surfactant in chemical polymerization reactions A phase-transfer catalyst for the quantitative determination of sulfide in the whole blood sample by extractive alkylation technique using GC-MS

Specifications

Melting Point 112°C to 116°C
Molecular Formula C12H16BNO4
MDL Number MFCD02179437
Quantity 5 g
Synonym 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)nitrobenzene
Molecular Weight (g/mol) 249.07 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.