Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Benzalkonium Chloride Solution NF (50%)
SDP

Catalog No. 14185388 Shop All Medisca Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-185-388 500 mL
14-185-288 100 mL
14-185-452 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-185-388 Supplier Medisca Supplier No. 026408
Add to Cart
Add to Cart

Benzalkonium Chloride Solution NF is a clear liquid; colorless or slightly yellow unless a color has been added. It has an aromatic odor.

Benzalkonium Chloride Solution NF is a clear liquid; colorless or slightly yellow unless a color has been added. It has an aromatic odor.

Specifications

Chemical Name or Material Benzalkonium Chloride Solution NF (50%)
Packaging Bottle, Amber Glass, Boston Round, 500mL, 38-439
Type Drug Standard
Molecular Weight (g/mol) Unknown
Quantity 500 mL
Grade NF
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.