Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzamide, 2-(3-aminopropoxy)-5-chloro-N-(1,1-dioxidobenzo[b]thien-6-yl)-hydrochloride | 2415263-08-8 | 98.7% | 429.32 | C18H18Cl2N2O4S | 5MG
SDP

Catalog No. 5000236424
Encompass_Preferred

HJC0416 hydrochloride is the hydrochloride salt of a small-molecule STAT3 inhibitor that reduces STAT3 phosphorylation, inhibits cell proliferation in vitro, and has shown antitumor activity in preclinical models. It is supplied as a white to off-white solid with high analytical purity and is intended for laboratory research use only.

  • Potent STAT3 inhibitor with reported in vitro IC50 0.04-1.97 μM.
  • Demonstrated oral bioavailability and antitumor activity in preclinical models.
  • High purity (98.7% by HPLC).
  • Molecular weight 429.32 and chemical formula C18H18Cl2N2O4S.
  • White to off-white solid appearance.
  • Storage: -20 °C, protect from light; in solvent: -80 °C (6 months) or -20 °C (1 month).
  • For laboratory research use only; not for human or clinical use.

Catalog No. 50-002-36424 Supplier Medchemexpress LLC Supplier No. HY12352A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HJC0416 hydrochloride is the hydrochloride salt of a small-molecule STAT3 inhibitor that reduces STAT3 phosphorylation, inhibits cell proliferation in vitro, and has shown antitumor activity in preclinical models. It is supplied as a white to off-white solid with high analytical purity and is intended for laboratory research use only.

  • Potent STAT3 inhibitor with reported in vitro IC50 0.04-1.97 μM.
  • Demonstrated oral bioavailability and antitumor activity in preclinical models.
  • High purity (98.7% by HPLC).
  • Molecular weight 429.32 and chemical formula C18H18Cl2N2O4S.
  • White to off-white solid appearance.
  • Storage: -20 °C, protect from light; in solvent: -80 °C (6 months) or -20 °C (1 month).
  • For laboratory research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.