Learn More
Medchemexpress LLC Benzamide, 2,6-dichloro-4-(cyclopentylamino)-N-[3-(1,1-dimethylethyl)-4-hydroxyphenyl] hydrochloride | 1660153-21-8 | 100.0% | 457.82 g/mol | C22H27Cl3N2O2 | 10 MG
SDP
Catalog No. 5000285336
Encompass_Preferred
FXR Antagonist 2 (hydrochloride) is a diarylamide-derived farnesoid X receptor (FXR) antagonist provided as the hydrochloride salt for research use. It is intended for biochemical and cellular studies of FXR-related signaling and metabolic-disease models. The compound is characterized by a molecular formula C22H27Cl3N2O2, molecular weight 457.82 g/mol, and high HPLC purity as reported on the certificate of analysis.
  • High purity suitable for research applications.
  • Hydrochloride salt form for improved solubility.
  • Diarylamide scaffold useful for FXR antagonism studies.
  • Supplied with a certificate of analysis including molecular weight and formula.
  • Available in small research pack sizes for assay development.
Catalog No. 50-002-85336 Supplier Medchemexpress LLC Supplier No. HY148874A10MG
FXR Antagonist 2 (hydrochloride) is a diarylamide-derived farnesoid X receptor (FXR) antagonist provided as the hydrochloride salt for research use. It is intended for biochemical and cellular studies of FXR-related signaling and metabolic-disease models. The compound is characterized by a molecular formula C22H27Cl3N2O2, molecular weight 457.82 g/mol, and high HPLC purity as reported on the certificate of analysis.
  • High purity suitable for research applications.
  • Hydrochloride salt form for improved solubility.
  • Diarylamide scaffold useful for FXR antagonism studies.
  • Supplied with a certificate of analysis including molecular weight and formula.
  • Available in small research pack sizes for assay development.