Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzamide, 3-amino | 3544-24-9 | 100.0% | 1 ML
SDP

Catalog No. 5000232380
Encompass_Preferred

3-Aminobenzamide (PARP-IN-1) is a small-molecule inhibitor of poly(ADP-ribose) polymerase (PARP) used in biochemical and cellular research to probe DNA repair and cell death pathways. It exhibits an IC50 of approximately 50 nM in CHO cells and is supplied as a high-purity reagent suitable for research applications.

  • Potent PARP inhibitor with an IC50 of approximately 50 nM in CHO cells.
  • High purity: HPLC 99.97%.
  • Provided as a concentrated solution (10 mM, 1 mL) or as a solid for flexibility in assays.
  • Chemical formula C7H8N2O; molecular weight 136.15 g/mol.

Catalog No. 50-002-32380 Supplier Medchemexpress LLC Supplier No. HY1202210MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

3-Aminobenzamide (PARP-IN-1) is a small-molecule inhibitor of poly(ADP-ribose) polymerase (PARP) used in biochemical and cellular research to probe DNA repair and cell death pathways. It exhibits an IC50 of approximately 50 nM in CHO cells and is supplied as a high-purity reagent suitable for research applications.

  • Potent PARP inhibitor with an IC50 of approximately 50 nM in CHO cells.
  • High purity: HPLC 99.97%.
  • Provided as a concentrated solution (10 mM, 1 mL) or as a solid for flexibility in assays.
  • Chemical formula C7H8N2O; molecular weight 136.15 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.