Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzamide, 3-amino- | 3544-24-9 | 100.0% | 500 MG
SDP

Supplier:  Medchemexpress LLC HY12022500MG

Encompass_Preferred

3-Aminobenzamide is a small-molecule PARP inhibitor used as a research reagent for biochemical and cell-based studies. It inhibits poly-ADP-ribose polymerase with reported IC50 ≈ 50 nM in CHO cells and is commonly used to investigate oxidant-induced myocyte dysfunction during reperfusion.

  • High purity as measured by HPLC.
  • Molecular weight 136.15 g/mol.
  • Chemical formula C7H8N2O.
  • CAS number 3544-24-9.
  • Appearance: off-white to light brown solid.
  • Stable at room temperature for up to 3 years; in solvent: -80°C (2 years), -20°C (1 year).
  • Suitable for biochemical and cell-based assays.

Catalog No. 50-002-33450


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.