Learn More
Medchemexpress LLC Benzamide, 4-[3-[4-cyano-3-(trifluoromethyl)phenyl]-5,5-dimethyl-4-oxo-2-thioxo-1-imidazolidinyl]-2-fluoro-N-(methyl-d3)- | 1443331-82-5 | >99.8% | C21H13D3F4N4O2S | 5 MG
SDP
Catalog No. 5000433638
Encompass_Preferred
Deutenzalutamide-d3 (Enzalutamide-d3) is a deuterium-labeled variant of Enzalutamide. Enzalutamide is recognized as an androgen receptor (AR) antagonist, demonstrating an IC50 of 36 nM in LNCaP prostate cells. This compound is primarily used for research purposes.
  • Deuterium-labeled compound for research applications
  • Potent androgen receptor antagonist
  • IC50 of 36 nM in LNCaP prostate cells
  • High purity with >99.8% by LCMS
Catalog No. 50-004-33638 Supplier Medchemexpress LLC Supplier No. HY70002S5MG
Deutenzalutamide-d3 (Enzalutamide-d3) is a deuterium-labeled variant of Enzalutamide. Enzalutamide is recognized as an androgen receptor (AR) antagonist, demonstrating an IC50 of 36 nM in LNCaP prostate cells. This compound is primarily used for research purposes.
  • Deuterium-labeled compound for research applications
  • Potent androgen receptor antagonist
  • IC50 of 36 nM in LNCaP prostate cells
  • High purity with >99.8% by LCMS