Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzamide, 4-[[4-[1-[3-(cyanomethyl)-1-(ethylsulfonyl)-3-azetidinyl]-1H-pyrazol-4-yl]-7H-pyrrolo... | 2789678-92-6 | 99.3% | C41H46N12O5S | 10MG
SDP

Catalog No. 5000259560
Encompass_Preferred

SJ10542 is a potent, selective PROTAC degrader that promotes degradation of JAK2 and JAK3 in cell-based assays, demonstrating low-nanomolar DC50 values and antitumor activity in preclinical models. Supplied as a high-purity solid for research use.

  • Potent degradation of JAK2 and JAK3 with low-nanomolar DC50 values.
  • High purity (HPLC 99.3%), suitable for biochemical and cellular studies.
  • White to off-white solid form, easy to handle and store.
  • Soluble in DMSO (46.67 mg/mL), enabling preparation of concentrated stock solutions.
  • Recommended storage: sealed at 4°C; in solution, store at -80°C for long-term stability.
  • Intended for research use in hematological malignancy and immunology studies.

Catalog No. 50-002-59560 Supplier Medchemexpress LLC Supplier No. HY14569610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SJ10542 is a potent, selective PROTAC degrader that promotes degradation of JAK2 and JAK3 in cell-based assays, demonstrating low-nanomolar DC50 values and antitumor activity in preclinical models. Supplied as a high-purity solid for research use.

  • Potent degradation of JAK2 and JAK3 with low-nanomolar DC50 values.
  • High purity (HPLC 99.3%), suitable for biochemical and cellular studies.
  • White to off-white solid form, easy to handle and store.
  • Soluble in DMSO (46.67 mg/mL), enabling preparation of concentrated stock solutions.
  • Recommended storage: sealed at 4°C; in solution, store at -80°C for long-term stability.
  • Intended for research use in hematological malignancy and immunology studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.