Learn More
Medchemexpress LLC Benzamide, 4-butoxy-N-[1,4-dihydro-2-(4-hydroxyphenyl)-4-oxo-3(2H)-quinazolinyl]- | 871100-12-8 | 99.8% | 431.48 Da | C25H25N3O4 | 25 MG
SDP
Catalog No. 5000284944
Encompass_Preferred
Quin-C7 is an orally active antagonist of the formyl peptide receptor 2 (FPR2/ALX) with reported anti-inflammatory activity in DSS-induced colitis models. Supplied as a high-purity research reagent, it is provided in solid form and as a DMSO solution for preclinical studies.
  • Orally active FPR2/ALX antagonist
  • Demonstrated anti-inflammatory effect in DSS-induced colitis models
  • High purity suitable for preclinical research
  • Supplied as powder and 10 mM solution in DMSO
  • Accompanied by datasheet and certificate of analysis
Catalog No. 50-002-84944 Supplier Medchemexpress LLC Supplier No. HYW66877525MG
Quin-C7 is an orally active antagonist of the formyl peptide receptor 2 (FPR2/ALX) with reported anti-inflammatory activity in DSS-induced colitis models. Supplied as a high-purity research reagent, it is provided in solid form and as a DMSO solution for preclinical studies.
  • Orally active FPR2/ALX antagonist
  • Demonstrated anti-inflammatory effect in DSS-induced colitis models
  • High purity suitable for preclinical research
  • Supplied as powder and 10 mM solution in DMSO
  • Accompanied by datasheet and certificate of analysis