Learn More
Medchemexpress LLC Benzamide, 4-butoxy-N-(2,4-difluorophenyl)- | 433967-28-3 | C17H17F2NO2 | 25 MG
SDP
Catalog No. 5000374894
Encompass_Preferred
VU 0357121 is a positive and highly selective mGlu5R allosteric modulator (PAM) with an EC50 of 33 nM. It is inactive or exhibits very weak antagonizing effects at other mGlu receptor subtypes.
  • Highly selective mGlu5R allosteric modulator (PAM)
  • EC50 of 33 nM
  • Inactive or very weakly antagonizing at other mGlu receptor subtypes
  • Enhances glutamate sensitivity of mGlu5
  • Does not bind at the MPEP allosteric site of mGlu5
  • Used for research purposes only
Catalog No. 50-003-74894 Supplier Medchemexpress LLC Supplier No. 25MGHY15393
VU 0357121 is a positive and highly selective mGlu5R allosteric modulator (PAM) with an EC50 of 33 nM. It is inactive or exhibits very weak antagonizing effects at other mGlu receptor subtypes.
  • Highly selective mGlu5R allosteric modulator (PAM)
  • EC50 of 33 nM
  • Inactive or very weakly antagonizing at other mGlu receptor subtypes
  • Enhances glutamate sensitivity of mGlu5
  • Does not bind at the MPEP allosteric site of mGlu5
  • Used for research purposes only