Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzamide, 4-butoxy-N-(2,4-difluorophenyl)- | 433967-28-3 | C17H17F2NO2 | 50 MG
SDP

Catalog No. 5000376114
Encompass_Preferred

VU 0357121 is a positive and highly selective mGlu5R allosteric modulator (PAM) with an EC50 of 33 nM. It is inactive or very weakly antagonizing at other mGlu receptor subtypes.

  • Positive and highly selective mGlu5R allosteric modulator
  • EC50 of 33 nM for mGlu5R
  • Inactive or very weakly antagonizing at other mGlu receptor subtypes
  • Appearance: solid, white to off-white
  • Solubility in DMSO: ≥ 50 mg/mL (163.76 mM)

Catalog No. 50-003-76114 Supplier Medchemexpress LLC Supplier No. HY1539350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

VU 0357121 is a positive and highly selective mGlu5R allosteric modulator (PAM) with an EC50 of 33 nM. It is inactive or very weakly antagonizing at other mGlu receptor subtypes.

  • Positive and highly selective mGlu5R allosteric modulator
  • EC50 of 33 nM for mGlu5R
  • Inactive or very weakly antagonizing at other mGlu receptor subtypes
  • Appearance: solid, white to off-white
  • Solubility in DMSO: ≥ 50 mg/mL (163.76 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.