Learn More
Medchemexpress LLC Benzamide, 4-fluoro-N-[2-[4-(7-methoxy-1-naphthalenyl)-1-piperazinyl]ethyl]-,hydrochloride | 286369-38-8 | 99.9% | 443.94 | 25 MG
SDP
Catalog No. 5000347625
Encompass_Preferred
S-14506 hydrochloride is a potent 5-HT1A agonist and a 5-HT2A/2C antagonist. It also exhibits dopamine antagonist properties by blocking dopamine D2 receptors. This compound inhibits the in vivo binding of [3H]raclopride in striatum and olfactory bulbs, with potential for research as an anxiolytic agent.
  • Potent 5-HT1A agonist.
  • 5-HT2A/2C antagonist.
  • Displays dopamine antagonist properties by blocking dopamine D2 receptors.
  • Inhibits the in vivo binding of [3H]raclopride in striatum and olfactory bulbs.
  • Potential for the research of anxiolytic agents.
Catalog No. 50-003-47625 Supplier Medchemexpress LLC Supplier No. 25MGHY110024
S-14506 hydrochloride is a potent 5-HT1A agonist and a 5-HT2A/2C antagonist. It also exhibits dopamine antagonist properties by blocking dopamine D2 receptors. This compound inhibits the in vivo binding of [3H]raclopride in striatum and olfactory bulbs, with potential for research as an anxiolytic agent.
  • Potent 5-HT1A agonist.
  • 5-HT2A/2C antagonist.
  • Displays dopamine antagonist properties by blocking dopamine D2 receptors.
  • Inhibits the in vivo binding of [3H]raclopride in striatum and olfactory bulbs.
  • Potential for the research of anxiolytic agents.