Learn More
Medchemexpress LLC Benzamide, 4-iodo-N-[2-[4-(2-methoxyphenyl)-1-piperazinyl]ethyl]-N-2-pyridinyl-, HCl | 220643-77-6 | 99.8% | 50 MG
SDP
Catalog No. 5000230118
Encompass_Preferred
p-MPPI hydrochloride is a selective 5-HT1A receptor antagonist used in preclinical pharmacology to study serotonin receptor function. It is provided as the hydrochloride salt, is reported to cross the blood-brain barrier, and is supplied at high purity for in vitro and in vivo research.
  • selective 5-HT1A receptor antagonist for pharmacological research
  • hydrochloride salt that crosses the blood-brain barrier
  • high purity (99.8%) with molecular weight 578.87
  • available as solid and 10 mM solution formats
  • suitable for in vitro and in vivo preclinical studies
Catalog No. 50-002-30118 Supplier Medchemexpress LLC Supplier No. HY12073850MG
p-MPPI hydrochloride is a selective 5-HT1A receptor antagonist used in preclinical pharmacology to study serotonin receptor function. It is provided as the hydrochloride salt, is reported to cross the blood-brain barrier, and is supplied at high purity for in vitro and in vivo research.
  • selective 5-HT1A receptor antagonist for pharmacological research
  • hydrochloride salt that crosses the blood-brain barrier
  • high purity (99.8%) with molecular weight 578.87
  • available as solid and 10 mM solution formats
  • suitable for in vitro and in vivo preclinical studies