Learn More
Medchemexpress LLC Benzamide, 4-iodo-N-[2-[4-(2-methoxyphenyl)-1-piperazinyl]ethyl]-N-2-pyridinyl hydrochloride | 220643-77-6 | 99.8% | 100 MG
SDP
Catalog No. 5000234877
Encompass_Preferred
p-MPPI hydrochloride is a selective 5-HT1A receptor antagonist used in research on serotonin receptor pharmacology; it is supplied as the monohydrochloride salt and crosses the blood-brain barrier.
  • Selective 5-HT1A receptor antagonist.
  • High assay purity by LCMS.
  • Supplied as a monohydrochloride salt for improved handling.
  • Molecular weight 578.87 and molecular formula C25H28ClIN4O2.
  • Suitable for receptor binding and behavioral pharmacology studies.
Catalog No. 50-002-34877 Supplier Medchemexpress LLC Supplier No. HY120738100MG
p-MPPI hydrochloride is a selective 5-HT1A receptor antagonist used in research on serotonin receptor pharmacology; it is supplied as the monohydrochloride salt and crosses the blood-brain barrier.
  • Selective 5-HT1A receptor antagonist.
  • High assay purity by LCMS.
  • Supplied as a monohydrochloride salt for improved handling.
  • Molecular weight 578.87 and molecular formula C25H28ClIN4O2.
  • Suitable for receptor binding and behavioral pharmacology studies.