Learn More
Medchemexpress LLC Benzamide, 5-(3-azetidinylamino)-N-[(1R)-1-[3-[5-[[[(1S,3R)-3-hydroxycyclopentyl)amino]methyl]-2- | 2817811-16-6 | C29H36N4O2S | 200 MG
SDP
Catalog No. 5000375640
Encompass_Preferred
XR8-89 is a potent papain-like protease (PLpro) inhibitor with an IC50 value of 0.1 μM. This compound induces conformational changes in SARS-CoV-2 papain-like protease, effectively inhibiting SARS-CoV-2 replication, making it suitable for research applications.
  • Potent papain-like protease (PLpro) inhibitor
  • IC50 value of 0.1 μM against papain-like protease
  • Induces conformational changes in SARS-CoV-2 papain-like protease
  • Inhibits SARS-CoV-2 replication
  • For SARS-CoV-2 research use
  • Purity of 97.13%
  • Appears as a light yellow to yellow solid
Catalog No. 50-003-75640 Supplier Medchemexpress LLC Supplier No. HY150784200MG
XR8-89 is a potent papain-like protease (PLpro) inhibitor with an IC50 value of 0.1 μM. This compound induces conformational changes in SARS-CoV-2 papain-like protease, effectively inhibiting SARS-CoV-2 replication, making it suitable for research applications.
  • Potent papain-like protease (PLpro) inhibitor
  • IC50 value of 0.1 μM against papain-like protease
  • Induces conformational changes in SARS-CoV-2 papain-like protease
  • Inhibits SARS-CoV-2 replication
  • For SARS-CoV-2 research use
  • Purity of 97.13%
  • Appears as a light yellow to yellow solid