Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzamide, 5-chloro-4-[(3,4-dichlorophenoxy)methyl]-2-fluoro-N-(methylsulfonyl)- | 1355631-24-1 | 99.4% | 426.67 | 1 ML
SDP

Catalog No. 5000351056
Encompass_Preferred

Nav1.7 inhibitor (compound II) is a potent sulfonamide Nav1.7 inhibitor. It has potential for treating various pain disorders, including acute, inflammatory, and neuropathic pain.

  • Potent Nav1.7 inhibitor
  • Purity 99.4%
  • Molecular weight 426.67
  • CAS number 1355631-24-1
  • Formula C15H11Cl3FNO4S
  • Appearance is solid, white to off-white
  • Shipping at room temperature in continental US; may vary elsewhere
  • Storage for powder at -20°C for 3 years, 4°C for 2 years
  • Storage for in solvent at -80°C for 2 years, -20°C for 1 year
  • Solubility (in vitro): ≥ 100 mg/mL (234.37 mM) in DMSO
  • Solubility (in vivo): ≥ 2.5 mg/mL (5.86 mM) in 10% DMSO and 90% corn oil
  • Target Nav1.7

Catalog No. 50-003-51056 Supplier Medchemexpress LLC Supplier No. HY1398510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nav1.7 inhibitor (compound II) is a potent sulfonamide Nav1.7 inhibitor. It has potential for treating various pain disorders, including acute, inflammatory, and neuropathic pain.

  • Potent Nav1.7 inhibitor
  • Purity 99.4%
  • Molecular weight 426.67
  • CAS number 1355631-24-1
  • Formula C15H11Cl3FNO4S
  • Appearance is solid, white to off-white
  • Shipping at room temperature in continental US; may vary elsewhere
  • Storage for powder at -20°C for 3 years, 4°C for 2 years
  • Storage for in solvent at -80°C for 2 years, -20°C for 1 year
  • Solubility (in vitro): ≥ 100 mg/mL (234.37 mM) in DMSO
  • Solubility (in vivo): ≥ 2.5 mg/mL (5.86 mM) in 10% DMSO and 90% corn oil
  • Target Nav1.7

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.