Learn More
Medchemexpress LLC Benzamide | 55-21-0 | MFCD00007968 | 1mL
SDP
Catalog No. 5000266469
Encompass_Preferred
Benzamide (Benzenecarboxamide) is a potent poly(ADP-ribose) polymerase (PARP) inhibitor Benzamide has protective activity against both glutamate- and methamphetamine (METH)-induced neurotoxicity in vitro Benzamide can attenuate the METH-induced dopamine depletions and exhibits neuroprotective activity in mice also has no acute effect on striatal dopamine metabolism and does not reduce body temperature[1]
Catalog No. 50-002-66469 Supplier Medchemexpress LLC Supplier No. HYZ028310MM/1ML
Benzamide (Benzenecarboxamide) is a potent poly(ADP-ribose) polymerase (PARP) inhibitor Benzamide has protective activity against both glutamate- and methamphetamine (METH)-induced neurotoxicity in vitro Benzamide can attenuate the METH-induced dopamine depletions and exhibits neuroprotective activity in mice also has no acute effect on striatal dopamine metabolism and does not reduce body temperature[1]