Learn More
Medchemexpress LLC Benzamide, n-[1-[[(cyanomethyl)amino]carbonyl]cyclohexyl]-4-[2-(4-methyl-1-piperazinyl)-4-thiazolyl]- | 294623-49-7 | MFCD18086856 | 99.9% | 466.60 g/mol | C24H30N6O2S | 10 MG
SDP
Catalog No. 5000244539
Encompass_Preferred
L-006235 is a potent, selective, reversible, and orally active inhibitor of cathepsin K (CAS 294623-49-7) used in research to study bone resorption and collagen degradation. It exhibits nanomolar activity in biological assays and is supplied as a solid or as a DMSO solution for analytical and pharmacology workflows.
  • Potent cathepsin K inhibition (IC50 ~5 nM)
  • High selectivity versus other cathepsins
  • Available as solid and 10 mM DMSO solution
  • High purity suitable for research applications
  • Stable when stored frozen; follow solvent storage guidelines
Catalog No. 50-002-44539 Supplier Medchemexpress LLC Supplier No. HY10335210MG
L-006235 is a potent, selective, reversible, and orally active inhibitor of cathepsin K (CAS 294623-49-7) used in research to study bone resorption and collagen degradation. It exhibits nanomolar activity in biological assays and is supplied as a solid or as a DMSO solution for analytical and pharmacology workflows.
  • Potent cathepsin K inhibition (IC50 ~5 nM)
  • High selectivity versus other cathepsins
  • Available as solid and 10 mM DMSO solution
  • High purity suitable for research applications
  • Stable when stored frozen; follow solvent storage guidelines