Learn More
Medchemexpress LLC Benzamide, N-[(1R)-2-[(trans-4-aminocyclohexyl)amino]-1-[(4-cyanophenyl)methyl]ethyl]- | 98.1% | 654.24 | 10 MG
SDP
Catalog No. 5000436781
Encompass_Preferred
trans-BAY-850 is the trans isomer of BAY-850, a potent and isoform selective ATPase family AAA domain-containing protein 2 (ATAD2) inhibitor, with an IC50 of 166 nM. This high purity compound is intended for research use only.
  • Trans isomer of BAY-850.
  • Potent and isoform selective ATPase family AAA domain-containing protein 2 (ATAD2) inhibitor.
  • IC50 of 166 nM.
  • Intended for research use only.
  • High purity of 98.12%.
  • Available in various quantities, including 5 mg, 10 mg, 25 mg, 50 mg, and 100 mg.
  • Solubility information is provided for both *In Vitro* and *In Vivo* experimental applications.
Catalog No. 50-004-36781 Supplier Medchemexpress LLC Supplier No. HY119254A10MG
trans-BAY-850 is the trans isomer of BAY-850, a potent and isoform selective ATPase family AAA domain-containing protein 2 (ATAD2) inhibitor, with an IC50 of 166 nM. This high purity compound is intended for research use only.
  • Trans isomer of BAY-850.
  • Potent and isoform selective ATPase family AAA domain-containing protein 2 (ATAD2) inhibitor.
  • IC50 of 166 nM.
  • Intended for research use only.
  • High purity of 98.12%.
  • Available in various quantities, including 5 mg, 10 mg, 25 mg, 50 mg, and 100 mg.
  • Solubility information is provided for both *In Vitro* and *In Vivo* experimental applications.