Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzamide, N-[(1S)-1-(aminocarbonyl)-4-[(2-chloro-1-iminoethyl)amino]butyl]-, hydrochloride | 1373232-26-8 | MFCD22201084 | 99.4% | 347.25 g/mol | C14H20Cl2N4O2 | 50 MG
SDP

Catalog No. 5000212924
Encompass_Preferred

Cl-amidine hydrochloride is an orally active peptidylarginine deiminase (PAD) inhibitor used in biochemical and cellular research to inhibit PAD-mediated protein citrullination, induce apoptosis, and investigate inflammation-related pathways. It is supplied as the hydrochloride salt and is intended for research use.

  • Orally active PAD inhibitor with reported activity against PAD1, PAD3, and PAD4.
  • High purity (99.4% by LCMS, batch COA).
  • Molecular formula C14H20Cl2N4O2; molecular weight 347.25 g/mol.
  • Store solid at -20°C under nitrogen; in solution store at -80°C for up to 6 months.
  • Provided in small research pack sizes, such as 50 MG.
  • Used to study protein citrullination, apoptosis, and inflammatory disease models.

Catalog No. 50-002-12924 Supplier Medchemexpress LLC Supplier No. HY100574A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cl-amidine hydrochloride is an orally active peptidylarginine deiminase (PAD) inhibitor used in biochemical and cellular research to inhibit PAD-mediated protein citrullination, induce apoptosis, and investigate inflammation-related pathways. It is supplied as the hydrochloride salt and is intended for research use.

  • Orally active PAD inhibitor with reported activity against PAD1, PAD3, and PAD4.
  • High purity (99.4% by LCMS, batch COA).
  • Molecular formula C14H20Cl2N4O2; molecular weight 347.25 g/mol.
  • Store solid at -20°C under nitrogen; in solution store at -80°C for up to 6 months.
  • Provided in small research pack sizes, such as 50 MG.
  • Used to study protein citrullination, apoptosis, and inflammatory disease models.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.