Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzamide, N-(3S)-1-azabicyclo[2.2.2]oct-3-yl-2,5-dimethoxy-, 2,2,2-trifluoroacetate (1:1) | 1336913-03-1 | 99.85% | 404.38 | 10 MG
SDP

Catalog No. 5000438351
Encompass_Preferred

PSEM 89S TFA is a selective and brain penetrant agonist for the resulting ion channels. It is orthogonally selective for Q79G and L141F, respectively. It is for research use only and not sold to patients.

  • Selective and brain penetrant agonist.
  • Orthogonally selective for Q79G and L141F.
  • Activates layer 2/3 cortical neurons expressing PSAML141F,Y115F-5HT3 high conductance (HC) at 10 or 30 μM.
  • Reversibly silenced transfected neurons by reducing cellular input resistance at 10 μM.
  • Strongly reduces photostimulation-evoked feeding in mice expressing PSAML141F,Y115F-glycine receptor (GlyR) (30 mg/kg).
  • Shows good brain penetrance in mice after minimally invasive intraperitoneal administration (30 mg/kg).

Catalog No. 50-004-38351 Supplier Medchemexpress LLC Supplier No. HY112217A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PSEM 89S TFA is a selective and brain penetrant agonist for the resulting ion channels. It is orthogonally selective for Q79G and L141F, respectively. It is for research use only and not sold to patients.

  • Selective and brain penetrant agonist.
  • Orthogonally selective for Q79G and L141F.
  • Activates layer 2/3 cortical neurons expressing PSAML141F,Y115F-5HT3 high conductance (HC) at 10 or 30 μM.
  • Reversibly silenced transfected neurons by reducing cellular input resistance at 10 μM.
  • Strongly reduces photostimulation-evoked feeding in mice expressing PSAML141F,Y115F-glycine receptor (GlyR) (30 mg/kg).
  • Shows good brain penetrance in mice after minimally invasive intraperitoneal administration (30 mg/kg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.