Learn More
Medchemexpress LLC Benzamide, N-(aminoiminomethyl)-2-methyl-5-(methylsulfonyl)-4-(1H-pyrrol-1-yl) hydrochloride | 211813-86-4 | MFCD31560490 | 99.5% | 356.83 | C14H17ClN4O3S | 10 MG
SDP
Catalog No. 5000243837
Encompass_Preferred
Eniporide hydrochloride is a small-molecule Na+/H+ exchanger inhibitor provided as the hydrochloride salt for use in biochemical and pharmacological research. It has been used in preclinical studies of cardiac ischemia/reperfusion and cellular ion regulation. The compound is supplied as a high-purity solid with recommended cold storage to preserve stability.
  • Potent Na+/H+ exchanger inhibitor
  • Hydrochloride salt form for improved stability
  • High purity (99.5%) suitable for research applications
  • Solid form, convenient for storage and handling
  • Recommended storage: 4°C; in solvent -80°C (6 months) or -20°C (1 month)
  • Suitable for studies of ion transport and cardiac models
Catalog No. 50-002-43837 Supplier Medchemexpress LLC Supplier No. HY106150B10MG
Eniporide hydrochloride is a small-molecule Na+/H+ exchanger inhibitor provided as the hydrochloride salt for use in biochemical and pharmacological research. It has been used in preclinical studies of cardiac ischemia/reperfusion and cellular ion regulation. The compound is supplied as a high-purity solid with recommended cold storage to preserve stability.
  • Potent Na+/H+ exchanger inhibitor
  • Hydrochloride salt form for improved stability
  • High purity (99.5%) suitable for research applications
  • Solid form, convenient for storage and handling
  • Recommended storage: 4°C; in solvent -80°C (6 months) or -20°C (1 month)
  • Suitable for studies of ion transport and cardiac models