Learn More
Medchemexpress LLC Benzamide, N-(aminoiminomethyl)-2-methyl-5-(methylsulfonyl)-4-(1H-pyrrol-1-yl) hydrochloride | 211813-86-4 | MFCD31560490 | 99.5% | 356.83 | C14H17ClN4O3S | 50 MG
SDP
Catalog No. 5000242725
Encompass_Preferred
Eniporide hydrochloride is a research-grade Na+/H+ exchange inhibitor provided as the hydrochloride salt for in vitro and in vivo studies of cellular pH regulation and myocardial ischemia/reperfusion. The reagent is characterized by high purity, defined solubility in common solvents, and recommended storage conditions to preserve stability for research use.
  • High purity suitable for research applications.
  • Available as solid and as solution in DMSO for experimental flexibility.
  • Soluble in DMSO and water with documented concentrations and handling notes.
  • Stable when stored under recommended sealed, low-temperature conditions.
  • Selective inhibition of Na+/H+ exchanger for mechanistic studies.
Catalog No. 50-002-42725 Supplier Medchemexpress LLC Supplier No. HY106150B50MG
Eniporide hydrochloride is a research-grade Na+/H+ exchange inhibitor provided as the hydrochloride salt for in vitro and in vivo studies of cellular pH regulation and myocardial ischemia/reperfusion. The reagent is characterized by high purity, defined solubility in common solvents, and recommended storage conditions to preserve stability for research use.
  • High purity suitable for research applications.
  • Available as solid and as solution in DMSO for experimental flexibility.
  • Soluble in DMSO and water with documented concentrations and handling notes.
  • Stable when stored under recommended sealed, low-temperature conditions.
  • Selective inhibition of Na+/H+ exchanger for mechanistic studies.