Learn More
Medchemexpress LLC Benzamide, n-hydroxy-3-[1-[(phenylthio)methyl]-1h-1,2,3-triazol-4-yl]- | 1316652-41-1 | MFCD21363002 | 99.3% | 326.37 g/mol | C16H14N4O2S | 10 MG
SDP
Catalog No. 5000227030
Encompass_Preferred
NCC-149 is a selective histone deacetylase 8 (HDAC8) inhibitor used in neural differentiation research. It is a small-molecule benzamide derivative (C16H14N4O2S; MW 326.37 g/mol; CAS 1316652-41-1) supplied as a solid with documented purity and batch COA.
  • Selective HDAC8 inhibitor useful for neural differentiation studies.
  • Batch-specific purity reported at 99.3% (COA).
  • Available as a small 10 MG solid suitable for research-scale experiments.
  • Solid stable at -20°C for up to 3 years; solvent solutions require colder storage.
  • Comes with COA, SDS, and product datasheet for quality and safety information.
Catalog No. 50-002-27030 Supplier Medchemexpress LLC Supplier No. HY11734810MG
NCC-149 is a selective histone deacetylase 8 (HDAC8) inhibitor used in neural differentiation research. It is a small-molecule benzamide derivative (C16H14N4O2S; MW 326.37 g/mol; CAS 1316652-41-1) supplied as a solid with documented purity and batch COA.
  • Selective HDAC8 inhibitor useful for neural differentiation studies.
  • Batch-specific purity reported at 99.3% (COA).
  • Available as a small 10 MG solid suitable for research-scale experiments.
  • Solid stable at -20°C for up to 3 years; solvent solutions require colder storage.
  • Comes with COA, SDS, and product datasheet for quality and safety information.