Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzamide, N-hydroxy-3,4-dimethoxy-N-methyl | 82461-57-2 | 99.8% | 211.21 g·mol⁻¹ | C10H13NO4 | 100MG
SDP

Catalog No. 5000239438
Encompass_Preferred

LK 614 is a membrane-permeable iron chelator provided for laboratory research. It is used in cell-based and biochemical studies to investigate protective effects against L-histidine-induced cell damage and other iron-dependent processes.

  • Membrane-permeable iron chelator.
  • Used to study protection against L-histidine-induced cell damage.
  • High purity (≈99.8%).
  • Molecular formula C10H13NO4.
  • Molecular weight 211.21 g·mol⁻¹.
  • Available in multiple package sizes (5 mg-500 mg).
  • Powder storage at -20°C; solutions stable at -80°C for 6 months and -20°C for 1 month.
  • Supplied as a solid powder for laboratory research use only.

Catalog No. 50-002-39438 Supplier Medchemexpress LLC Supplier No. HY126164100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LK 614 is a membrane-permeable iron chelator provided for laboratory research. It is used in cell-based and biochemical studies to investigate protective effects against L-histidine-induced cell damage and other iron-dependent processes.

  • Membrane-permeable iron chelator.
  • Used to study protection against L-histidine-induced cell damage.
  • High purity (≈99.8%).
  • Molecular formula C10H13NO4.
  • Molecular weight 211.21 g·mol⁻¹.
  • Available in multiple package sizes (5 mg-500 mg).
  • Powder storage at -20°C; solutions stable at -80°C for 6 months and -20°C for 1 month.
  • Supplied as a solid powder for laboratory research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.