Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Benzamidine

Catalog No. 1207250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-072-50G 50 g
12-072-10G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-072-50G Supplier Sigma Aldrich Supplier No. 1207250G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Benzamidine was used in the preparation of heparan sulfate-rich proteoglycan from mouse mammary epithelial cells
  • It has been used to prevent enzymatic degradation during the purification procedure of bovine factor xii (hageman factor).benzamidine is an inhibitor of plasmin and has been used in the radioimmunoassay of glucagon in human plasma
  • It prevents the destruction of thyrotropin-releasing hormone (trh) added to whole blood in vitro before radioimmunoassay of trh

Specifications

CAS 618-39-3
Molecular Formula C7H8N2
Linear Formula C6H5C(=NH)NH2
MDL Number MFCD00042826
Quantity 50 g
Molecular Weight (g/mol) 120.15
Percent Purity ≥95.0%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.