Learn More
Apexbio Technology LLC Benzamidine HCl 1670-14-0 1g
SDP
Catalog No. 5000089205
Encompass_Preferred
Benzamidine hydrochloride (CAS 1670-14-0) is a competitive inhibitor of serine proteases acting through reversible binding at the active sites of these enzymes It effectively inhibits multiple proteases including trypsin tryptase urokinase-type plasminogen activator (uPA) factor Xa thrombin and tissue-type plasminogen activator (tPA) Benzamidine HCl is widely utilized in studies investigating the coagulation and fibrinolytic systems as well as in physiological and pharmacological research focusing on serine protease activity
Catalog No. 50-000-89205 Supplier Apexbio Technology LLC Supplier No. C68661000
Add to Cart
Add to Cart
Benzamidine hydrochloride (CAS 1670-14-0) is a competitive inhibitor of serine proteases acting through reversible binding at the active sites of these enzymes It effectively inhibits multiple proteases including trypsin tryptase urokinase-type plasminogen activator (uPA) factor Xa thrombin and tissue-type plasminogen activator (tPA) Benzamidine HCl is widely utilized in studies investigating the coagulation and fibrinolytic systems as well as in physiological and pharmacological research focusing on serine protease activity