Learn More
Research Products International Corp Benzamidine Hydrochloride, 25 Grams
SDP
Catalog No. 50488531
Encompass_Preferred
Competitive serine protease inhibitor which forms stable complexes with and blocks the active sites of enzymes. Target enzymes include trypsin, hymotrypsin, kallikzein and plasmin.
Catalog No. 50-488-531 Supplier Research Products International Corp Supplier No. B1200025.0
Competitive serine protease inhibitor which forms stable complexes with and blocks the active sites of enzymes. Target enzymes include trypsin, hymotrypsin, kallikzein and plasmin.