Learn More
Medchemexpress LLC HY-W018781 500mg Medchemexpress, Benzamidine (hydrochloride) CAS:1670-14-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW018781500MG
Encompass_Preferred
Medchemexpress, HY-W018781 500mg Benzamidine (hydrochloride) CAS:1670-14-0 Benzamidine hydrochloride is an reversible competitive inhibitor of trypsin-like serine proteases, with Kis of 97 µM, 21 µM, 20 µM and 110 µM for uPA, trypsin, tryptase and factor Xa, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3430