Learn More
Alkali Scientific Benzamidine Hydrochloride, 100 G
SDP
Catalog No. 5000660680
Encompass_Preferred
Benzamidine Hydrochloride is a high-purity protease inhibitor provided in a 100g package, ideal for use in biochemical and molecular biology applications. As a potent inhibitor of serine proteases, it is frequently used to prevent enzymatic degradation in protein purification and enzyme activity assays. Benzamidine Hydrochloride is commonly employed in laboratory workflows to inhibit trypsin, thrombin, and other proteases during protein analysis, ensuring that protein samples remain intact and active. This reagent is widely used in cell culture, biochemical research, and the preparation of cell lysates and other protein-rich samples where protease activity must be suppressed.
Catalog No. 50-006-60680 Supplier Alkali Scientific Supplier No. BEN001CP
Benzamidine Hydrochloride is a high-purity protease inhibitor provided in a 100g package, ideal for use in biochemical and molecular biology applications. As a potent inhibitor of serine proteases, it is frequently used to prevent enzymatic degradation in protein purification and enzyme activity assays. Benzamidine Hydrochloride is commonly employed in laboratory workflows to inhibit trypsin, thrombin, and other proteases during protein analysis, ensuring that protein samples remain intact and active. This reagent is widely used in cell culture, biochemical research, and the preparation of cell lysates and other protein-rich samples where protease activity must be suppressed.