Learn More
Gold Biotechnology Inc Benzamidine Hydrochloride Monohydrate 100 g
SDP
Catalog No. NC2107475
Encompass
Benzamidine hydrochloride monohydrate is an aromatic amidine. It is used in protein extractions as a strong reversible inhibitor of serine proteases such as trypsin, thrombin, plasmin and other trypsin-like proteases. Compared to aprotinin, benzamidine is considered to be just as effective in preventing the degradation of glucogon.
This item is not returnable. View return policy
Catalog No. NC2107475 Supplier Gold Biotechnology Inc Supplier No. B050100
Benzamidine hydrochloride monohydrate is an aromatic amidine. It is used in protein extractions as a strong reversible inhibitor of serine proteases such as trypsin, thrombin, plasmin and other trypsin-like proteases. Compared to aprotinin, benzamidine is considered to be just as effective in preventing the degradation of glucogon.