Learn More
Alkali Scientific Benzamidine Hydrochloride, 25 G
Supplier: Alkali Scientific BEN002CP
Benzamidine Hydrochloride is a highly pure protease inhibitor offered in a 25g package, designed for laboratory use in biochemical and molecular biology studies. It is primarily utilized to inhibit serine proteases, such as trypsin and thrombin, to protect proteins from degradation during various analytical and preparatory procedures. This reagent is essential in experiments requiring the preservation of protein structure and activity, particularly in enzyme assays, protein purification, and cell lysate preparation. Benzamidine Hydrochloride is widely applied in research focused on enzyme kinetics, protein-protein interactions, and the study of protease functions.