Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzamidine Hydrochloride 96.0+%, TCI America™
SDP

Catalog No. B0013100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0013100G 100 g
B001325G 25 g
B0013500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0013100G Supplier TCI America Supplier No. B0013100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1670-14-0
Molecular Formula C7H9ClN2
Molecular Weight (g/mol) 156.613
MDL Number MFCD00013025
InChI Key LZCZIHQBSCVGRD-UHFFFAOYSA-N
Synonym benzamidine hydrochloride, benzimidamide hydrochloride, benzenecarboximidamide hydrochloride, benzenecarboximidamide, monohydrochloride, benzamidinium chloride, benzamidine hcl, benzamidine, hydrochloride, amidinobenzene hydrochloride, benzamidine, monohydrochloride, amidinobenzene hydrochloride 1:1
PubChem CID 80289
IUPAC Name benzenecarboximidamide;hydrochloride
SMILES C1=CC=C(C=C1)C(=N)N.Cl

Specifications

Melting Point 171°C
Color White
pH 4.0 to 6.5 (50g/L 25°C)
Quantity 100 g
Formula Weight 156.61
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzamidine Hydrochloride
RTECSNumber CV6260000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.