Learn More
Bioworld Benzamidine Separopore (Agarose) 6B Columns
Supplier:  Bioworld 201700242
Encompass
Applications: Benzamidine-Separopore 6B is p-aminobenzamidine covalently attached to Separopore 6B by the epoxy coupling method. p-Aminobenzamidine (PAB), isan inhibitor of trypsin-like serine protease. Trypsin and trypsin-like serine proteases bind to Benzamidine-Separopore6B and can thus be used for purification and/or removal of these substances.Benzamidine-Separopore6B can be used for purifying serine proteases such as trypsin, bovine thrombin, urokinase, human enterokinase, acrosin, native plasminogen, kallikrein, prekallikrein, collagenase and clostripain. For recombinant purification, Benzamidine-Separopore6B columns can be used for removal of serine proteases such as thrombin and enterokinase after cleavage of purification tags.
Catalog No. 50-197-643