Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzamidoxime, 95%

Catalog No. AAH2606803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2606803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2606803 Supplier Thermo Scientific Chemicals Supplier No. H2606803

missing translation for 'validMainframeMsg'

CAS: 613-92-3 | C7H8N2O | 136.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 613-92-3
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.15
MDL Number MFCD00031485 MFCD00474011
InChI Key MXOQNVMDKHLYCZ-UHFFFAOYSA-N
Synonym benzamidoxime, benzamide oxime, n'-hydroxybenzimidamide, benzohydroxamamide, n-hydroxybenzamidine, phenylhydroxamidine, n-hydroxybenzenecarboximidamide, ccris 2953, n'-hydroxybenzamidine, benzamidoxim
PubChem CID 7259353
ChEBI CHEBI:83354
IUPAC Name N'-hydroxybenzenecarboximidamide
SMILES N\C(=N/O)C1=CC=CC=C1

Specifications

Melting Point 72°C to 76°C
Quantity 1 g
Formula Weight 136.15
Percent Purity 95%
Chemical Name or Material Benzamidoxime
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 210-361-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.