Learn More
Medchemexpress LLC HY-B1546A 10mg Medchemexpress, Benzamil hydrochloride CAS:161804-20-2 Purity:>98%
SDP
Catalog No. 501932968
Encompass_Preferred
Medchemexpress, HY-B1546A 10mg Benzamil hydrochloride CAS:161804-20-2 Benzamil hydrochloride is a specific blocker of sodium channel ( ENaC ). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2968 Supplier Medchemexpress LLC Supplier No. HYB1546A10MG
Medchemexpress, HY-B1546A 10mg Benzamil hydrochloride CAS:161804-20-2 Benzamil hydrochloride is a specific blocker of sodium channel ( ENaC ). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.