Learn More
Medchemexpress LLC Benzamil hydrochloride | 161804-20-2 | 99.7% | 100 MG
SDP
Catalog No. 5000412111
Encompass_Preferred
Benzamil hydrochloride (Benzylamiloride hydrochloride), an Amiloride analogue, is a Na+/Ca2+ exchanger (NCX) inhibitor (IC50~100 nM). It is also a non-selective Deg/epithelial sodium channels (ENaC) blocker, and can potentiate myogenic vasoconstriction. Benzamil hydrochloride inhibits TRPP3-mediated Ca2+-activated currents, with an IC50 of 1.1 μM.
  • Na+/Ca2+ exchanger (NCX) inhibitor
  • Non-selective Deg/epithelial sodium channels (ENaC) blocker
  • Can potentiate myogenic vasoconstriction
  • Inhibits TRPP3-mediated Ca2+-activated currents
Catalog No. 50-004-12111 Supplier Medchemexpress LLC Supplier No. HYB1546A100MG
Benzamil hydrochloride (Benzylamiloride hydrochloride), an Amiloride analogue, is a Na+/Ca2+ exchanger (NCX) inhibitor (IC50~100 nM). It is also a non-selective Deg/epithelial sodium channels (ENaC) blocker, and can potentiate myogenic vasoconstriction. Benzamil hydrochloride inhibits TRPP3-mediated Ca2+-activated currents, with an IC50 of 1.1 μM.
  • Na+/Ca2+ exchanger (NCX) inhibitor
  • Non-selective Deg/epithelial sodium channels (ENaC) blocker
  • Can potentiate myogenic vasoconstriction
  • Inhibits TRPP3-mediated Ca2+-activated currents