Learn More
Medchemexpress LLC Benzamil hydrochloride | 161804-20-2 | 99.7% | 50 MG
SDP
Catalog No. 5000411479
Encompass_Preferred
Benzamil hydrochloride is an Amiloride analogue that acts as a Na+/Ca2+ exchanger (NCX) inhibitor (IC50~100 nM). It also functions as a non-selective Deg/epithelial sodium channels (ENaC) blocker and can enhance myogenic vasoconstriction. This product inhibits TRPP3-mediated Ca2+-activated currents with an IC50 of 1.1 μM.
  • Inhibits neuronal and heterologously expressed small conductance Ca2+-activated K+ channels.
  • Shown to improve survival in stroke-prone spontaneously hypertensive rats (SHRSP) when treated at 0.7 mg/kg/day (s.c.).
Catalog No. 50-004-11479 Supplier Medchemexpress LLC Supplier No. HYB1546A50MG
Benzamil hydrochloride is an Amiloride analogue that acts as a Na+/Ca2+ exchanger (NCX) inhibitor (IC50~100 nM). It also functions as a non-selective Deg/epithelial sodium channels (ENaC) blocker and can enhance myogenic vasoconstriction. This product inhibits TRPP3-mediated Ca2+-activated currents with an IC50 of 1.1 μM.
  • Inhibits neuronal and heterologously expressed small conductance Ca2+-activated K+ channels.
  • Shown to improve survival in stroke-prone spontaneously hypertensive rats (SHRSP) when treated at 0.7 mg/kg/day (s.c.).