Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzanilide 98%, ACROS Organics™

Click to view available options
Quantity:
5g
50g
250g
1kg
Packaging:
Glass bottle
Plastic bottle
93-98-1
C13H11NO
197.24
MFCD00003069
ZVSKZLHKADLHSD-UHFFFAOYSA-N
benzanilide,n-benzoylaniline,benzamide, n-phenyl,n-phenyl-benzamide,benzoic acid anilide,unii-ak1b12366o,phenyl-n-benzamide,benzanilid,benzoylanilide,n-benzoyl aniline
7168
N-phenylbenzamide
C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2
93-98-1
C13H11NO
197.24
MFCD00003069
ZVSKZLHKADLHSD-UHFFFAOYSA-N
benzanilide,n-benzoylaniline,benzamide, n-phenyl,n-phenyl-benzamide,benzoic acid anilide,unii-ak1b12366o,phenyl-n-benzamide,benzanilid,benzoylanilide,n-benzoyl aniline
7168
N-phenylbenzamide
C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2
Boiling Point 117°C (10.0mmHg)
Quantity 5g
Color Gray to Green-Yellow
Melting Point 161°C to 165°C
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C6H5CONHC6H5
Beilstein 12, 262
Merck Index 15, 1063
Solubility Information Solubility in water: insoluble. Other solubilities: 1g/60 mL alcohol,1g/7 mL boiling alcohol,slightly soluble in ether,acetic acid
Formula Weight 197.24
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material Benzanilide, 98%
Show More Show Less

EINECSNumber : 202-292-7

TSCA : TSCA