Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzanthrone 99%, Thermo Scientific™

Catalog No. AC105271000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC105271000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC105271000 Supplier Thermo Scientific Chemicals Supplier No. 105271000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82-05-3
Molecular Formula C17H10O
Molecular Weight (g/mol) 230.266
InChI Key HUKPVYBUJRAUAG-UHFFFAOYSA-N
Synonym benzanthrone, 7h-benz de anthracen-7-one, benzanthrenone, ms-benzanthrone, dye, benzanthrone, benzoanthrone, mesobenzanthrone, 7h-benzo de anthracen-7-one, 1,9-benzanthrone
PubChem CID 6697
IUPAC Name benzo[a]phenalen-7-one
SMILES C1=CC=C2C(=C1)C3=CC=CC4=C3C(=CC=C4)C2=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 168°C to 172°C
Color Khaki to Yellow
Infrared Spectrum Authentic
Quantity 100 g
Beilstein 07, 518
Merck Index 15, 1065
Formula Weight 230.27
Percent Purity ≥98.5%
Physical Form Fine Powder
Chemical Name or Material Benzanthrone, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.